ENA369381836 Enamine

2-chloro-1-(4-fluoro-3-methylphenyl)ethan-1-one

  • MDL number MFCD00129465
  • Cas Number : 696616-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101068

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101068
Availability details for
Quantity
  • Availability
  • Add to Cart