ENA369381837 Enamine

2-methoxypropanethioamide

  • MDL number MFCD16077951
  • Cas Number : 1250027-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58...60
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101069

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101069
Availability details for
Quantity
  • Availability
  • Add to Cart