ENA369381838 Enamine

ethyl 2-(aminomethyl)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD09904789
  • Cas Number : 120237-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101070

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101070
Availability details for
Quantity
  • Availability
  • Add to Cart