ENA369381841 Enamine

4-(3-methylphenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD16779596
  • Cas Number : 22899-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101074
Availability details for
Quantity
  • Availability
  • Add to Cart