ENA369381844 Enamine

2-(1-methoxyethyl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD21873189
  • Cas Number : 955400-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101077

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101077
Availability details for
Quantity
  • Availability
  • Add to Cart