ENA369381850 Enamine

5-({[2-(dimethylamino)ethyl](methyl)amino}methyl)pyridin-2-amine

  • MDL number MFCD18868422
  • Cas Number : 1197332-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100106
Availability details for
Quantity
  • Availability
  • Add to Cart