ENA369381853 Enamine

1-amino-N-(cyclopropylmethyl)cyclopentane-1-carboxamide

  • MDL number MFCD12162200
  • Cas Number : 1183226-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100110

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100110
Availability details for
Quantity
  • Availability
  • Add to Cart