ENA369381869 Enamine

N-(3-chloropropyl)cyclopentanecarboxamide

  • MDL number MFCD08715097
  • Cas Number : 925591-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100011
Availability details for
Quantity
  • Availability
  • Add to Cart