ENA369381881 Enamine

3-(3-chloro-4-fluorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD11190937
  • Cas Number : 1152540-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101192

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101192
Availability details for
Quantity
  • Availability
  • Add to Cart