ENA369381884 Enamine

3-(4H-1,2,4-triazol-3-yl)propanoic acid

  • MDL number MFCD14641027
  • Cas Number : 844439-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101197
Availability details for
Quantity
  • Availability
  • Add to Cart