ENA369381886 Enamine

2-(5-chlorothiophen-2-yl)propanoic acid

  • MDL number MFCD14529722
  • Cas Number : 1215995-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101200
Availability details for
Quantity
  • Availability
  • Add to Cart