ENA369381902 Enamine

5-cyclopropyl-1-ethyl-1H-pyrazol-3-amine

  • MDL number MFCD09834278
  • Cas Number : 1172529-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111...113
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101225

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101225
Availability details for
Quantity
  • Availability
  • Add to Cart