ENA369381903 Enamine

8-oxo-5,6,7,8-tetrahydronaphthalene-2-sulfonyl chloride

  • MDL number MFCD10691397
  • Cas Number : 1152559-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101226
Availability details for
Quantity
  • Availability
  • Add to Cart