ENA369381905 Enamine

3-(7-chloro-1H-1,3-benzodiazol-2-yl)propanoic acid

  • MDL number MFCD12150288
  • Cas Number : 1179224-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101228
Availability details for
Quantity
  • Availability
  • Add to Cart