ENA369381909 Enamine

N-benzyl-2,2-dimethylcyclopentan-1-amine

  • MDL number MFCD22056412
  • Cas Number : 1376297-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101233

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101233
Availability details for
Quantity
  • Availability
  • Add to Cart