ENA369381910 Enamine

4-[benzyl(methyl)amino]butan-1-ol

  • MDL number MFCD12186544
  • Cas Number : 98901-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101234

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101234
Availability details for
Quantity
  • Availability
  • Add to Cart