ENA369381911 Enamine

1-(3-methylbutyl)-1H-1,3-benzodiazol-2-amine hydrobromide

  • MDL number MFCD22056413
  • Cas Number : 1375969-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101235
Availability details for
Quantity
  • Availability
  • Add to Cart