ENA369381922 Enamine

2-[(7-chloroquinolin-4-yl)sulfanyl]propanoic acid

  • MDL number MFCD09941252
  • Cas Number : 874487-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101274
Availability details for
Quantity
  • Availability
  • Add to Cart