ENA369381923 Enamine

3-amino-1-(3-chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD14689754
  • Cas Number : 1250022-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101275
Availability details for
Quantity
  • Availability
  • Add to Cart