ENA369381925 Enamine

5-chloro-2-(2-methoxyethoxy)benzonitrile

  • MDL number MFCD16769702
  • Cas Number : 1250894-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-101277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101277
Availability details for
Quantity
  • Availability
  • Add to Cart