ENA369381932 Enamine

6-chloro-N-(3,4-dihydro-2H-1-benzopyran-4-yl)pyrimidin-4-amine

  • MDL number MFCD17293468
  • Cas Number : 1275835-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101285
Availability details for
Quantity
  • Availability
  • Add to Cart