ENA369381942 Enamine

N,N-dimethylpiperazine-2-carboxamide

  • MDL number MFCD18268340
  • Cas Number : 85817-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101297

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101297
Availability details for
Quantity
  • Availability
  • Add to Cart