ENA369381946 Enamine

ethyl 2-amino-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylate

  • MDL number MFCD01680496
  • Cas Number : 24237-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101307

Safety Information

signalword Warning
pcodes P264,P270,P280,P301+P312,P302+P352,P330,P332+P313,P362,P501
MSDS https://www.enaminestore.com/catalog/EN300-101307
pictograms GHS07
hstatements H302,H315
Availability details for
Quantity
  • Availability
  • Add to Cart