ENA369381948 Enamine

2-(cyanosulfanyl)-1-[2-(trifluoromethyl)phenyl]ethan-1-one

  • MDL number MFCD22056419
  • Cas Number : 947499-50-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101309

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101309
Availability details for
Quantity
  • Availability
  • Add to Cart