ENA369381959 Enamine

4-chloro-2-(4-methylpiperazin-1-yl)pyrimidine

  • MDL number MFCD14607614
  • Cas Number : 700803-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100168
Availability details for
Quantity
  • Availability
  • Add to Cart