ENA369381960 Enamine

4-aminocyclopent-2-ene-1-carboxylic acid hydrochloride

  • MDL number MFCD22056336
  • Cas Number : 1376003-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141...143
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100170

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100170
Availability details for
Quantity
  • Availability
  • Add to Cart