ENA369381970 Enamine

4-(3,4-dichlorophenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD16779594
  • Cas Number : 99817-29-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101353
Availability details for
Quantity
  • Availability
  • Add to Cart