ENA369381976 Enamine

ethyl 3-{3-[(methylcarbamoyl)methyl]-1,2,4-oxadiazol-5-yl}propanoate

  • MDL number MFCD22056424
  • Cas Number : 1376291-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-101361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101361
Availability details for
Quantity
  • Availability
  • Add to Cart