ENA369381988 Enamine

1-[2-(4-nitrophenyl)ethyl]-2,5-dihydro-1H-pyrrole-2,5-dione

  • MDL number MFCD13285256
  • Cas Number : 1292476-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101378
Availability details for
Quantity
  • Availability
  • Add to Cart