ENA369381995 Enamine

5-(azetidine-1-sulfonyl)-2-chlorobenzoic acid

  • MDL number MFCD09041674
  • Cas Number : 926227-65-8
  • Molecular Weight : 275.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-100014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100014
Availability details for
Quantity
  • Availability
  • Add to Cart