ENA369381996 Enamine

N-[(furan-2-yl)(phenyl)methylidene]hydroxylamine

  • MDL number MFCD00093276
  • Cas Number : 2067-52-9
  • Molecular Weight : 187.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-101386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101386
Availability details for
Quantity
  • Availability
  • Add to Cart