ENA369382006 Enamine

4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-2-methylbenzoic acid

  • MDL number MFCD22196402
  • Cas Number : 161291-72-1
  • Molecular Weight : 281.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-101426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101426
Availability details for
Quantity
  • Availability
  • Add to Cart