ENA369382007 Enamine

1-(3-chlorophenyl)pyrrolidin-3-amine

  • MDL number MFCD11894455
  • Cas Number : 1178748-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101427
Availability details for
Quantity
  • Availability
  • Add to Cart