ENA369382014 Enamine

2-(4-ethylphenyl)-2-methylpropan-1-amine

  • MDL number MFCD16301795
  • Cas Number : 1225815-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101438

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101438
Availability details for
Quantity
  • Availability
  • Add to Cart