ENA369382021 Enamine

1-acetylpyrrolidine-3-carboxylic acid

  • MDL number MFCD17272529
  • Cas Number : 712270-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145...147
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100180

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-100180
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart