ENA369382022 Enamine

3-[(2-carboxyethyl)carbamoyl]propanoic acid

  • MDL number MFCD00481984
  • Cas Number : 78238-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101465

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101465
Availability details for
Quantity
  • Availability
  • Add to Cart