ENA369382024 Enamine

3-(3-methoxypropoxy)aniline

  • MDL number MFCD11652535
  • Cas Number : 1023277-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-101467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101467
Availability details for
Quantity
  • Availability
  • Add to Cart