ENA369382042 Enamine

2,3-dihydro-1-benzofuran-3,6-diol

  • MDL number MFCD11149382
  • Cas Number : 1020947-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100184

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100184
Availability details for
Quantity
  • Availability
  • Add to Cart