ENA369382047 Enamine

3-methyl-1-(pyridin-2-ylmethyl)-1H-pyrazol-5-amine

  • MDL number MFCD16092662
  • Cas Number : 1224047-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101530

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101530
Availability details for
Quantity
  • Availability
  • Add to Cart