ENA369382049 Enamine

4-(3-methoxyphenyl)-1,2,5-oxadiazol-3-amine

  • MDL number MFCD16779597
  • Cas Number : 1249939-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101532
Availability details for
Quantity
  • Availability
  • Add to Cart