ENA369382052 Enamine

3-(4,4-dimethyl-2,5-dioxoimidazolidin-1-yl)benzonitrile

  • MDL number MFCD13326127
  • Cas Number : 1306198-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101535

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101535
Availability details for
Quantity
  • Availability
  • Add to Cart