ENA369382054 Enamine

1-(1H-1,2,3,4-tetrazol-5-yl)butan-1-amine

  • MDL number MFCD12796321
  • Cas Number : 31603-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 277...279
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101537

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101537
Availability details for
Quantity
  • Availability
  • Add to Cart