ENA369382055 Enamine

potassium trifluoro(1-methyl-1H-pyrazol-4-yl)boranuide

  • MDL number MFCD09992927
  • Cas Number : 1365970-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100187
Availability details for
Quantity
  • Availability
  • Add to Cart