ENA369382056 Enamine

(3,4-dimethylphenyl)methanesulfonyl chloride

  • MDL number MFCD11052243
  • Cas Number : 1000350-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101543
Availability details for
Quantity
  • Availability
  • Add to Cart