ENA369382057 Enamine

N-cyclopropyl-2-(piperidin-4-yloxy)acetamide hydrochloride

  • MDL number MFCD08459588
  • Cas Number : 1260763-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183...185
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101544

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-101544
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart