ENA369382059 Enamine

1-[(1-aminobutan-2-yl)oxy]-3-(trifluoromethyl)benzene

  • MDL number MFCD16081676
  • Cas Number : 1247503-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101546

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101546
Availability details for
Quantity
  • Availability
  • Add to Cart