ENA369382063 Enamine

(4-methanesulfinylphenyl)methanamine hydrochloride

  • MDL number MFCD11100897
  • Cas Number : 1189719-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 217...219
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101556

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101556
Availability details for
Quantity
  • Availability
  • Add to Cart