ENA369382065 Enamine

3-(1-benzothiophen-3-yl)propanoic acid

  • MDL number MFCD11045734
  • Cas Number : 26461-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101558

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101558
Availability details for
Quantity
  • Availability
  • Add to Cart