ENA369382071 Enamine

4-methyl-1H,2H,3H,4H-pyrido[2,3-b]pyrazin-2-one

  • MDL number MFCD08062792
  • Cas Number : 1314974-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 161...163
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101566

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101566
Availability details for
Quantity
  • Availability
  • Add to Cart