ENA369382076 Enamine

2-amino-3-(3-chlorophenyl)propanoic acid hydrochloride

  • MDL number MFCD02065493
  • Cas Number : 120108-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101573

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101573
Availability details for
Quantity
  • Availability
  • Add to Cart